missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Basic Fuchsin, Electrophoresis Reagent

Product Code. 15427879
Change view
Click to view available options
Quantity:
5g
25g
100g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15427879 100g 100g
15417879 25g 25g
15407879 5g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15427879 Supplier Thermo Scientific™ Supplier No. J66494.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 632-99-5
Format Crystalling powder
Formula Weight 337.86
Melting Point 268° to 270°C (dec.)
Molecular Formula C20H20ClN3
Packaging Plastic bottle
Quantity 100g
Storage Requirements Ambient
Synonym C.I. 42510,Fuchsin basic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.