missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Basic Fuchsin, 88%, pure

Product Code. p-62241 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10455661

Brand: Thermo Scientific Chemicals 419141000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 632-99-5 | C20H20ClN3 | 337.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-99-5
Molecular Formula C20H20ClN3
Molecular Weight (g/mol) 337.85
MDL Number MFCD00012569
InChI Key AXDJCCTWPBKUKL-UHFFFAOYSA-N
Synonym Basic Violet 14, hydrochloride, C.I. 42510, Rosaniline chloride
PubChem CID 12447
IUPAC Name 4-[(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
SMILES [H+].[Cl-].CC1=CC(C=CC1=N)=C(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1

Specifications

Absorbance 1800 (at lambda min.)
Melting Point 250.0°C
CAS Min % 83.0
Absorption Ratio 1.1 to 1.4
CAS Max % 100.0
Dye Content 83% min. (on dry substance)
Assay Percent Range 88%
Beilstein 13, 765
Merck Index 15, 5715
Solubility Information Solubility in water: 4g/L (25°C). Other solubilities: soluble in alcohol and acids, practically insoluble in ether, soluble 30g/L in ethanol (25°C)
Infrared Spectrum Authentic
Lambda Max 549 to 555nm (in 50:50 IMS:water)
Formula Weight 337.84
Percent Purity 88%
Grade Pure
Packaging Glass bottle
Color Green
Quantity 100 g
Flash Point 200°C
Physical Form Fine Powder
Chemical Name or Material Basic Fuchsin, 88%
Show More Show Less
compliance-icons
Product Identifier
  • Basic Fuchsin
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Magenta

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.