missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Basic Fuchsin, 70+%, pure

Product Code. 10791894 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10791894 500 g Glass bottle 500g
10589082 25 g Glass bottle 25g
10035203 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10791894 Supplier Thermo Scientific Chemicals Supplier No. 401695000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 632-99-5 | C20H20ClN3 | 337.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-99-5
Molecular Formula C20H20ClN3
Molecular Weight (g/mol) 337.85
MDL Number MFCD00012569
InChI Key AXDJCCTWPBKUKL-UHFFFAOYSA-N
Synonym Basic Violet 14, hydrochloride, C.I. 42510, Rosaniline chloride
PubChem CID 12447
IUPAC Name 4-[(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
SMILES [H+].[Cl-].CC1=CC(C=CC1=N)=C(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1

Specifications

Melting Point 250.0°C
Absorption Ratio 1.16 to 1.35
Dye Content 70% min.
Beilstein 13, 765
Merck Index 15, 5715
Solubility Information Solubility in water: 4g/L (25°C). Other solubilities: soluble in alcohol and acids, practically insoluble in ether, soluble 30g/L in ethanol (25°C)
Infrared Spectrum Authentic
Lambda Max 547 to 552nm
Formula Weight 337.84
Percent Purity ≥70%
Grade Pure
Packaging Glass bottle
Density 200g/mL
Color Green
Quantity 500 g
Flash Point 200°C
Physical Form Glistening Crystalline Powder
Chemical Name or Material Basic Fuchsin, Pure
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.