missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Basic Fuchsin

Product Code. 11419803
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419803 5 g 5g
11429803 25 g 25g
11439803 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419803 Supplier Thermo Scientific Chemicals Supplier No. A12952.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 632-99-5 | C20H20ClN3 | 337.85 g/mol

Biological stain for distinguishing coli and aerogene bacteriaBasic Fuchsin is useful for Ziehl-Neelson staining to detect acid-fast bacilli. It is also used to stain tubercle bacillus and bacterial flagella. It plays an important role for tissue sections to visualize elastic fibers, cardiac and skeletal muscle tissue. It also acts as bacterial and histological stains. Further, it is used to stain cellular elements, certain tissues, bacteria and also acts as a local anti-infective.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biological stain for distinguishing coli and aerogene bacteriaBasic Fuchsin is useful for Ziehl-Neelson staining to detect acid-fast bacilli. It is also used to stain tubercle bacillus and bacterial flagella. It plays an important role for tissue sections to visualize elastic fibers, cardiac and skeletal muscle tissue. It also acts as bacterial and histological stains. Further, it is used to stain cellular elements, certain tissues, bacteria and also acts as a local anti-infective.

Solubility
Soluble in water, alcohol, diethyl ether and acids practice. Insoluble in ether.

Notes
Incompatible with oxidizing agents, reducing agents, acids, acid chlorides, acid anhydrides and chloroformates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-99-5
Molecular Formula C20H20ClN3
Molecular Weight (g/mol) 337.85
MDL Number MFCD00012569
InChI Key AXDJCCTWPBKUKL-UHFFFAOYSA-N
Synonym basic violet 14, fuchsin, fuchsin basic, magenta, rosaniline, fuchsine, basic fuchsine, basic magenta, rose aniline, fuchsin, basic
PubChem CID 12447
IUPAC Name 4-[(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
SMILES [H+].[Cl-].CC1=CC(C=CC1=N)=C(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1

Specifications

Melting Point 268°C to 270°C (decomposition)
Quantity 5 g
Beilstein 4166684
Merck Index 14,5652
Solubility Information Soluble in water,alcohol,diethyl ether and acids practice; Insoluble in ether.
Formula Weight 337.86
Chemical Name or Material Basic Fuchsin
compliance-icons
Product Identifier
  • Basic Fuchsin
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Magenta

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.