missing translation for 'onlineSavingsMsg'
Learn More

Labcold™ Basic Freezers

Product Code. 12328067
Click to view available options
Capacity (Metric):
70 L
505 L
Dimensions (D x W x H) Interior:
365 x 305 x 370 mm
545 x 645 x 1490 mm
Voltage:
230 V
240 V
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12328067

Brand: Labcold™ RLVL03203

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Providing a large capacity combined with good economical value for laboratory and general use. Labcold™ Basic Freezers are suitable for the storage of certain temperature-sensitive materials such as enzymes.

  • Door lock
  • CFC and HCFC free
  • Temperature range of -18° to -25°C
  • Analog temperature display
  • High temperature warning light
  • Operator-controlled defrost

Specifications

Type General Purpose Freezer
Capacity (Metric) 70 L
Temperature Range -18°C to -25°C
Door Style Solid
No. of Drawers 2
Dimensions (D x W x H) Interior 365 x 305 x 370 mm
Depth (Metric) Exterior 490 mm
Depth (Metric) Interior 365 mm
Height (Metric) Exterior 835 mm
Height (Metric) Interior 370 mm
Width (Metric) Exterior 500 mm
Width (Metric) Interior 305 mm
Voltage 240 V
Frequency 50/60 Hz
Weight (Metric) 28 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.