missing translation for 'onlineSavingsMsg'
Learn More

Basic Bouffant Cap

Product Code. 16662604
Change view
Click to view available options
Diameter (English):
21 in.
24 in.
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Unit Size
16662604 24 in. Pack of 1000
16652604 21 in. Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16662604 Supplier New source - PPE Supplier No. HMMC24

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Basic bouffant caps made of lightweight, comfortable spunbound polypropylene fabric, and are ideal for use in the laboratory or the wider working environment for promoting general hygiene.

  • General purpose disposable bouffant caps available in two sizes
  • Promotes general hygiene in the laboratory or wider workplace environment
  • With elasticated seams for an easy, comfortable fit
  • Non-woven, spunbound polypropylene fabric construction
  • Latex-free
  • Supplied as a carton of 10 bags, 100 bouffants per bag

Specifications

Product Type Bouffant cap
Material Spunbond Polypropylene
Color White
Disposable Single-use
Latex Free Yes
Sterility Non-sterile
Certifications/Compliance (Europe) PPE CAT I, ISO tested to ISO3801 & ISO13934-1. Complies with EC Directive 1935/2004/EC, Regulation (EU) No.10/2011, Regulation (EU) No.2016/1416 and Regulation (EU) 2017/752
Quantity 1000/Pk.
Diameter (English) 24 in.
Diameter (Metric) 60.9 cm
Weight (Metric) 10 gsm
For Use With (Application) General purpose in the lab or cleanroom
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.