missing translation for 'onlineSavingsMsg'
Learn More

Basic Blue 26, TRC

Product Code. 30008544
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008544 2.5 g 2.5g
30009644 500 mg 500mg
17900505 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008544 Supplier Toronto Research Chemicals Supplier No. TRCB1187482.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2580-56-5
Molecular Formula C33 H32 N3 . Cl
Molecular Weight (g/mol) 506.08
Synonym C.I. Basic Blue 26, N-[4-[[4-(Dimethylamino)phenyl][4-(phenylamino)-1-naphthalenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-methylmethanaminium chloride, Victoria Blue B, ADC Victoria Blue B, Aizen Victoria Blue BH, BTK Victoria Blue, Basazol C Blue 57, Basic Blue 26, Basic Blue B, Basic Victoria Blue B, Basonyl Blue 640, Basonyl Blue 644, Basovict Victoria Blue, C-WR Blue 8, C.I. 44045, Calcozine Blue B, Conbasic Blue AK, Dycosbasic Victoria Blue B, Flexo Blue 630, Flexo Blue 640, Hecto Blue B, Hidaco Victoria Blue B, Libbase Victoria Blue LB, Lowacryl Blue 26, Mitsui Victoria Blue B, Ravi Victoria Blue B, Tertrophene Blue, Victoria Blue, Victoria Blue 2B, Victoria Blue B 353, Victoria Blue B chloride, Victoria Blue BA, Victoria Blue BH, Victoria Blue BN, Victoria Blue BN CI 44045, Victoria Blue BP, Victoria Blue BS, Victoria Blue BSA, Victoria Blue BX, Victoria Blue FB, Victoria Pure Blue B, Victoria Pure Blue BC
IUPAC Name [4-[(4-anilinonaphthalen-1-yl)-[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
SMILES [Cl-].CN(C)c1ccc(cc1)C(=C2C=CC(=[N+](C)C)C=C2)c3ccc(Nc4ccccc4)c5ccccc35

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C33H31N3.ClH/c1-35(2)27-18-14-24(15-19-27)33(25-16-20-28(21-17-25)36(3)4)31-22-23-32(30-13-9-8-12-29(30)31)34-26-10-6-5-7-11-26;/h5-23H,1-4H3;1H
Formula Weight 505.2285
Chemical Name or Material Victoria Blue B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.