missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Basic Benchtop pH Meters

Product Code. 11672220
Click to view available options
Dimensions (L x W x H):
182 L x 240 D x 74 mm H
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11672220

Brand: Hanna Instruments™ HI2210

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Basic, affordable pH benchtop meter that features automatic calibration to 1 or 2 points with a choice of 5 preprogrammed buffers.

  • High specification pH meter offering outstanding value for money
  • Designed with an extra wide LCD screen which can display pH/mV and temperature readings
  • Readings are automatically compensated for temperature to ensure accuracy
  • One or two calibration points
  • Up to five memorised calibration values
  • Supplied with refillable glass pH electrode, temperature probe, electrode stand, calibration sachets, electrode refill solution and mains power lead

Specifications

Accuracy (Temperature) ±0.4°C
Accuracy (pH) ±0.01
Calibration Automatic
Range (pH) -2 to 16
Resolution (pH) 0.01
Calibration Points 1 or 2 with 5 memorized buffer values
Temperature Range (Metric) -20.0°C to +120.0°C
Resolution (Temperature) 0.1°C
Temperature Probe Calibration HI 7662 stainless steel temperature probe with 1 m (3.3’) cable (included)
Display Type LCD
Inputs BNC
Dimensions (L x W x H) 182 L x 240 D x 74 mm H
Material Glass Body
Model HI2210-02
Power Supply 230V
Probe Type HI 7662 with 1 m (3.3') cable (included)
Detection Range -2 to 16 pH
Temperature Compensation Automatic or manual from –9.9 to 120°C
Temperature (Metric) Operating -20.0°C to +120.0°C
Type Basic Bench Top pH Meter
Length (Metric) 182 mm
Height (Metric) 74 mm
Depth (Metric) 240 mm
Electrical Requirements 240V, 50/60Hz
Voltage 240 V
Frequency 50 Hz
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.