missing translation for 'onlineSavingsMsg'
Learn More

Base Number (BN) Standard: 40 mg KOH/g in Hydrocarbon Oil, VHG™

Product Code. 18092020
Change view
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18092020 100 g 100g
17929959 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18092020 Supplier VHG Labs Supplier No. VHGBN40100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Base Number (BN) Standard: 40 mg KOH/g in Hydrocarbon Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Base Number
Analyte or Component Names Base Number
Shipping Condition Room Temperature
Concentration 40 mg KOH/g
Solvent or Matrix Hydrocarbon Oil
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.