Learn More
Base/Neutrals Surrogate Standard, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep CLP90-SB
Code : 04
Additional Details : CAS Number : 2199-69-1 Weight : 0.10000kg
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
1,2-Dichlorobenzene-d4 | 2199-69-1 | 1000μg/mL |
2-Fluorobiphenyl | 321-60-8 | 1000μg/mL |
Nitrobenzene-d5 | 4165-60-0 | 1000μg/mL |
p-Terphenyl-d14 | 1718-51-0 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Base/Neutrals Surrogate Standard | |
1 mL | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
1000μg/mL | |
MeCl:Acetone |
2mL amber prescored glass ampule | |
2199-69-1,321-60-8,4165-60-0,1718-51-0 | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
1,2-Dichlorobenzene-d4,2-Fluorobiphenyl |
Safety and Handling
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : dichloromethane, acetone, nitrobenzene
Product intended for laboratory use only