Learn More
Descripción
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| 1,2-Dichlorobenzene-d4 | 2199-69-1 | 1000μg/mL |
| 2-Fluorobiphenyl | 321-60-8 | 1000μg/mL |
| Nitrobenzene-d5 | 4165-60-0 | 1000μg/mL |
| p-Terphenyl-d14 | 1718-51-0 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Especificaciones
Especificaciones
| Chemical Name or Material | Base/Neutrals Surrogate Standard |
| Packaging | 5mL amber prescored glass ampule |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity. |
| CAS | 2199-69-1, 321-60-8, 4165-60-0, 1718-51-0 |
| Analyte or Component Names | 1,2-Dichlorobenzene-d4,2-Fluorobiphenyl |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Element | 1,2-Dichlorobenzene-d4,2-Fluorobiphenyl |
| Solvent or Matrix | MeCl:Acetone |
| Quantity | 5 mL |
| Mostrar más |
Seguridad y manipulación
- Base/Neutrals Surrogate Standard, 5 mL
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H361-Suspected of damaging fertility or the unborn child.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: dichloromethane, acetone, nitrobenzene-d5
Product intended for laboratory use only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.