missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Basal cell cytokeratin Antibody (RCK103), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97729AF750
This item is not returnable.
View return policy
Description
Basal cell cytokeratin Monoclonal specifically detects Basal cell cytokeratin in Human, Rat, Porcine, Avian, Canine, Chicken, Guinea Pig, Hamster, Rabbit, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| Basal cell cytokeratin | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| RCK103 is a mouse monoclonal IgG1 antibody derived by fusion of SP2/0-Ag14 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with a mix of cell preparations containing Human Cytokeratins. | |
| 0.1 mL | |
| This monoclonal antibody stains basal cells in combined and stratified epithelial tissues. It recognizes the stem cell population, including the so-called amplifying cells in the prostate epithelium. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| RCK103 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| Mouse | |
| Protein A or G purified | |
| Primary | |
| Human, Rat, Pig, Avian, Canine, Chicken, Guinea Pig, Hamster, Rabbit, Zebrafish | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction