missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Basal Bodies of Cillia Antibody (LhS 28), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-50114CL1
This item is not returnable.
View return policy
Description
Basal Bodies of Cillia Monoclonal antibody specifically detects Basal Bodies of Cillia in Hamster,Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Immunocytochemistry/Immunofluorescence
Specifications
| Basal Bodies of Cillia | |
| Monoclonal | |
| CoraFluor 1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Hamster, Human | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Immunocytochemistry/Immunofluorescence | |
| LhS 28 | |
| PBS | |
| BHK a21 Cytoskeletal preparation | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction