missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BarX1 Recombinant Rabbit Monoclonal Antibody (K01_2L50)
Description
BarX1 Recombinant Monoclonal Antibody for Western Blot
BarX1 is a member of the Bar subclass of homeobox transcription factors. Studies of the mouse and chick homolog suggest the encoded protein may play a role in developing teeth and craniofacial mesenchyme of neural crest origin. The protein may also be associated with differentiation of stomach epithelia.
Specifications
Specifications
| Antigen | BarX1 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_2L50 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4 |
| Gene | BARX1 |
| Gene Accession No. | Q9HBU1 |
| Gene Alias | BarH-like homeobox 1; BARX homeobox 1; BARX1; Homeobox protein BarH-like 1; LOW QUALITY PROTEIN: homeobox protein BarH-like 1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?