missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BarX1 Antibody (S01-3A3), Novus Biologicals™
Rabbit Monoclonal Antibody
£328.00 - £500.00
Specifications
| Antigen | BarX1 |
|---|---|
| Clone | S01-3A3 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625373
|
Novus Biologicals
NBP3-19835-50ul |
50 μg |
£328.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18606933
|
Novus Biologicals
NBP3-19835-100ul |
100 μg |
£500.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BarX1 Monoclonal antibody specifically detects BarX1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| BarX1 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 56033 | |
| IgG | |
| Affinity purified |
| S01-3A3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| BarH-like homeobox 1, BARX homeobox 1, homeobox protein BarH-like 1 | |
| A synthetic peptide of human BarX1 (Uniprot # Q9HBU1) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title