missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Barnstead™ Pretreatment for Point of Use Water Purification Systems

Product Code. 15920216
Change view
Click to view available options
Cartridge Type:
Pretreatment Cartridge
Unit Size:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cartridge Type Unit Size
15920216 Pretreatment Cartridge 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15920216 Supplier Thermo Scientific™ Supplier No. 50157886
  • ONLINE EXCLUSIVE PRICE
    £1260.00
    £1230.70 / Pack of 1
    Save £29.30
    2% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Barnstead Pretreatment for Point of Use Water Purification Systems

Barnstead Pretreatment contains PreFilter, Hardness Stabilizer and Carbon Filter. The unit contains pressure regulator and pressure gauges. Space saving design comes with Wall Mount capability.

  • Use pretreatment when feed water quality doesn't match the requirements for your system
  • Contains pre-filter, hardness stabilizer and carbon filter
TRUSTED_SUSTAINABILITY

Specifications

Cartridge Type Pretreatment Cartridge
Description Pretreatment includes Particle Filter, Carbon Filter and Hardness Stabilizer Catridges ( D502113, D502114 and D502115 )
For Use With (Equipment) Smart2Pure Pro
Includes Pressure Regulator, Pressure Gauges, Three Cartridges and Wall Mount
Dimensions (L x W x H) 14.5 in x 13.3 in x 20.0 in.
Weight (English) 24.5 lb.
Weight (Metric) 11.4 kg
Product Type Water Purification System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.