Learn More
Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables
Shop All Thermo Scientific ProductsDescription
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables. Choose the cartridge pack according to your feed water and system. Cartridge packs ship with 0.2 micron final filter. Other products include UV lamp and low water level pump protector.
- Simple, one-connection installation and removal
Includes: One 0.2μm absolute final filter
Specifications
Specifications
| For Use With (Equipment) | Nanopure Water Purifications System |
| Type | Deionized Water Feed (Low Organics, Type I Water) |