Learn More
Thermo Scientific™ Barnstead™ LabTower™ TII Water Purification System
Convert tap water into high-purity water with the Thermo Scientific™ Barnstead™ LabTower™ TII Water Purification System, a complete solution integrating purification and tank.
Brand: Thermo Scientific™ 50132196
Code : A0
Additional Details : Weight : 0.80000kg
Description
Convert tap water into high-purity water with the Thermo Scientific™ Barnstead™ LabTower™ TII Water Purification System, a complete solution integrating purification and tank. This high-tech system is designed to continuously recirculate purified reservoir water to preserve water quality even during periods of inactivity. Conserve bench space with this stand-alone design which includes an integrated 100L reservoir. Castors allow for easy relocation within the lab.
Consistent water quality:
- Type 2 laboratory water meets ASTM Type II, CSLS-CLRW and ISO 3696 international standards.
- The LabTower is designed to continuously recirculate purified reservoir water to preserve water quality even during periods of inactivity.
Compact mobile design:
- Three system options are available with permeate flows of 20, 40 or 60L/hr.
- Stand-alone design with integrated 100L reservoir takes up zero bench space.
- Castors allow easy relocation.
- Systems can be upgraded later to accommodate growing water demands.
Two ways to dispense water:
- Dispense directly from the system via the dispensing valve for calibration solutions, reagent preparation, filling of containers, general rinsing purposes, etc.
- Dispense from the reservoir to supply autoclaves, dishwashers, ultrapure water systems, etc.
Integrated 100L reservoir:
- Up to 100L of high purity water is ready for dispensing when needed.
- High purity polypropylene reservoir is opaque to light.
- Conical bottom allows complete emptying for efficient cleaning and disinfection.
- Adjustable setting for reservoir volume can be programmed for times when demand is low, allowing you to only store water that you can use.
Clear information on system display:
- The large four-line display is code-protected to prevent accidental changes in system settings.
- Illuminated and easily readable, the display provides information on resistivity or conductivity and operating mode status, such as “roduction”, “stand-by” or sanitization and volume of the reservoir in percent.
GLP-compliant documentation:
- LabTower TII has been developed to meet or exceed GLP requirements.
- Recorded and traceable data can be obtained by print-out via the RS-232 interface and accessory printer.
- Highly qualified and precise measurement of the conductivity is ensured by the cell constant of 0.01 cm-1 to an accuracy of ±0.1°C.
Pretreatment:
- 5μm prefilter and activated carbon in the combi-filter cartridge protect the RO membrane from free chlorine and particles.
- Hardness stabilizing cartridge protects the reverse osmosis stage from hard water.
Reverse osmosis and reservoir:
- Pretreated water is pressure-forced through the permeable reverse osmosis module removing 97- 99% of all inorganic ions and 99% of dissolved organic substances, as well as microorganisms and particles.
- The built-in 100L reservoir has a conical bottom outlet to optimize cleaning and sanitization.
Ion exchange and UV:
- RO membrane removes up to 99% of impurities. All remaining ions are removed by the low TOC, high-purity resins in the ion exchange cartridge.
- Irradiation with UV light (TII units with UV option only) eliminates any bacteria or germs that are present.
Specifications
99% | |
Barnstead LabTower TII | |
3/4 in. NPT | |
Tap | |
Type 2 | |
No | |
1 to 6 bar | |
<1 | |
2°C to 35°C | |
1500 mm | |
66 kg | |
100/240 V |
0.1 to 1.0μS/cm | |
<1 CFU/mL | |
No | |
LabTower TII 40 with 3 L/min. pump | |
10 to 1 Mohm.cm | |
Water Purification System | |
2 to 6 bar | |
40 L/hr. at 15°C | |
450 mm | |
580 mm | |
100/240 V 50/60 Hz | |
50/60 Hz |