Learn More
Thermo Scientific™ Barnstead™ Easypure™ II
This product has been discontinued. For additional information or alternate products please contact your sales representative.
Brand: Thermo Scientific™ D8611
Code : 03
Additional Details : Weight : 0.50000kg
Description
This product has been discontinued. For additional information or alternate products please contact your sales representative. The recommended replacement for this product is Thermo Scientific™ Barnstead™ MicroPure™.
Features:- Each system produces ultrapure water that exceeds the quantitative specifications of Type 1 water as described in ASTM™ D1193, ISO 3696, and CLSI-CLRW.
- Three cartridges in series provide increased capacity over similar systems.
- A 0.2μm final filter at the point of use provides water free of particles and bacteria.
- Quiet pump provides optimal flowrates and silent operation.
- Standby mode recirculates purified water during inactivity to ensure purity and allows for on-demand use.
- All components are constructed of inert materials, ensuring maximum ionic and organic purity.
- Supplied wall bracket allows increased flexibility when choosing a location.
- Integral low water and low pressure pump protectors protect the pump in the event feedwater is interrupted to the system.
Easypure II:
- Base unit
- For buffer prep, titration, solutions, AA
- TOC 5 to 10ppb
Easypure II UV:
- Ultralow organics
- For HPLC, TOC, GC/MS
- TOC 1 to 5ppb
Easypure II UFI:
- Eliminates pyrogens
- For cell and tissue culture, monoclonal antibody production, electrophoresis
- TOC 5 to 10ppb
Easypure II UV:
- Lowest levels of organics, pyrogens and nucleases
- For PCR 2-D electrophoresis, cell culture, blotting
- TOC 1 to 5ppb
Ordering Information:
Optional accessories include Remote Dispenser (0903515) and replacement consumables (UV lamps, ultrafilters and cleaning syringe); order separately. Contact your Sales Representative for details.
Includes:
Wall mounting bracket, cleaning syringe, UV lamp (Easypure II UV and UV/UF only), ultrafilter (Easypure II UF and UV/UF only).
Requires:
- Cartridges and filters
Recommended for:
- HPLC
- ICP
- GC/MS
- Cell and tissue culture
- PCR
Note:
- Feedwater must be pre-treated by reverse osmosis, distillation or deionization
Specifications
CE, CSA | |
Discontinued; replacement product is 50132370 | |
0.001 EU/mL | |
Pretreated | |
Type 1 | |
No | |
Water Purification System | |
4°C to 40°C | |
<1 μM/mL | |
100/240 V |
0.055 µS/cm | |
<1 CFU/mL | |
No | |
Up to 1 L/min. | |
18.2 Mohm.cm at 25°C | |
1 to 5 ppb | |
100 psig (6.9 bar) | |
UV/UF model | |
100/240 V 50/60 Hz | |
50/60 Hz |