Learn More
Thermo Scientific™ Barnstead™ E-Pure™ Optional Accessories
Product Code. 11486348 Shop All Thermo Scientific Products
Click to view available options
Type:
Low Water Level Pump Protector
Remote Dispenser
Voltage:
0/1 V
240 V
Unit Size:
Each
Product Code. 11486348
Brand: Thermo Scientific™ D8952
Only null left
Add to basket
Add to basket
Optimize your ultrapure water purification system with Thermo Scientific™ Barnstead™ E-Pure™ Optional Accessories, including a remote dispenser.
Optimize your ultrapure water purification system with Thermo Scientific™ Barnstead™ E-Pure™ Optional Accessories. Increase flexibility in your lab with the remote dispenser, which delivers water up to 8' (2.4m) from the system and ships with a mounting bracket. Gain protection with the low pressure pump protector which alerts the E-Pure of an inadequate feed water condition in a pressurized feed line. The low water level pump protector alerts the system of an inadequate feed water condition when fed from a reservoir.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Barnstead E-Pure
Type Remote Dispenser
Voltage 0/1 V