Learn More
Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits
Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
E-Pure 3-holder
E-Pure 3-holder, Organic free
E-Pure 4-holder
E-Pure 4-holder, Organic free
E-Pure Ultrapure Water Purification System
Feedwater Source:
N/A
Pretreated
Pretreated feed
Tap
Unit Size:
Each
Pack of 3
Description
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
Specifications
Specifications
| Description | Cartridge kit for E-Pure 4-holder, Tap feed, Organic free |
| Feedwater Source | Tap |
| For Use With (Equipment) | E-Pure 4-holder, Organic free |
| Type | Cartridge Kit |