Learn More
Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits.
Brand: Thermo Scientific™ D50228
Code : 03
Additional Details : Weight : 6.46200kg
Description
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.Specifications
Cartridge Kit for E-Pure 4-holder; Pretreated feed, Organic free | |
Pretreated | |
Cartridge Kit for E-Pure 4-holder; Pretreated feed, Organic free | |
Cartridge Kit |