missing translation for 'onlineSavingsMsg'
Learn More

Barnidipine hydrochloride, MedChemExpress

Product Code. 16475076
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475076 100 mg 100mg
16474108 10 mg 10mg
16440467 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16475076 Supplier Medchem Express Supplier No. HY107322
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Barnidipine hydrochloride (Mepirodipine hydrochloride) is an L-type calcium antagonist (CaA) with high affinity for [3H] initrendipine binding sites (Ki=0.21 nmol/l), has selective action against CaA receptors. Barnidipine hydrochloride (Mepirodipine hydrochloride) is an antihypertensive drug and acts by the reduction of peripheral vascular resistance secondary to its vasodilatory action.

Barnidipine hydrochloride (Mepirodipine hydrochloride) is an L-type calcium antagonist (CaA) with high affinity for [3H] initrendipine binding sites (Ki=0.21 nmol/l), has selective action against CaA receptors.

Specifications

Chemical Name or Material Barnidipine hydrochloride
CAS 104757-53-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H30ClN3O6
Quantity 100 mg
Synonym Mepirodipine hydrochloride YM-09730-5
Solubility Information DMSO : 250 mg/mL (473.48 mM; Need ultrasonic)
SMILES O=C(C1=C(C)NC(C)=C(C(OC)=O)[C@@H]1C2=CC([N+]([O-])=O)=CC=C2)O[C@@H]3CN(CC4=CC=CC=C4)CC3.[H]Cl
Molecular Weight (g/mol) 528.0
Formula Weight 528.0
Percent Purity 98.77%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.