missing translation for 'onlineSavingsMsg'
Learn More

Barium zirconium oxide, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11371559
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11371559

Brand: Thermo Scientific Alfa Aesar 014028.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Barium zirconium oxide is involved in the preparation of its nano powder, which finds application in the gas sensing performance of thick films especially with ammonia gas. Copper(II) oxide doping with yttrium-doped barium zirconate is used as an electrolyte in solid oxide fuel cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12009-21-1
Molecular Formula BaO3Zr
Molecular Weight (g/mol) 276.548
MDL Number MFCD00014189
InChI Key DQBAOWPVHRWLJC-UHFFFAOYSA-N
Synonym barium zirconate, barium 2+ oxozirconiumbis olate, ba.o3zr, barium 2+ ;dioxido oxo zirconium, barium zirconate, powder, <10 mum, barium zirconate, nanopowder, <50 nm particle size trace metals basis
PubChem CID 16212444
IUPAC Name barium(2+);dioxido(oxo)zirconium
SMILES [O-][Zr](=O)[O-].[Ba+2]

Specifications

Melting Point 2500°C
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 99% (Metal basis)
Solubility Information Insoluble in water.
Formula Weight 276.53
Odor Odorless
Appearance Off-white powder
Density 5.52 g/cm3
Chemical Name or Material Barium zirconium oxide
Show More Show Less
compliance-icons
Product Identifier
  • Barium zirconium oxide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Barium zirconate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.