missing translation for 'onlineSavingsMsg'
Learn More

Barium zirconium oxide, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11361559
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361559 1 kg 1kg
11371559 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11361559 Supplier Thermo Scientific Alfa Aesar Supplier No. 014028.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Barium zirconium oxide is involved in the preparation of its nano powder, which finds application in the gas sensing performance of thick films especially with ammonia gas. Copper(II) oxide doping with yttrium-doped barium zirconate is used as an electrolyte in solid oxide fuel cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12009-21-1
Molecular Formula BaO3Zr
Molecular Weight (g/mol) 276.548
MDL Number MFCD00014189
InChI Key DQBAOWPVHRWLJC-UHFFFAOYSA-N
Synonym barium zirconate, barium 2+ oxozirconiumbis olate, ba.o3zr, barium 2+ ;dioxido oxo zirconium, barium zirconate, powder, <10 mum, barium zirconate, nanopowder, <50 nm particle size trace metals basis
PubChem CID 16212444
IUPAC Name barium(2+);dioxido(oxo)zirconium
SMILES [O-][Zr](=O)[O-].[Ba+2]

Specifications

Melting Point 2500°C
Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99% (Metal basis)
Solubility Information Insoluble in water.
Formula Weight 276.53
Odor Odorless
Appearance Off-white powder
Density 5.52
Chemical Name or Material Barium zirconium oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.