missing translation for 'onlineSavingsMsg'
Learn More

Barium titanium oxide, polymeric precursor, (Oxide ^= 35 wt%)

Product Code. 11384727
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
Product Code. Quantity unitSize
11384727 50 g 50g
11394727 2 g 2g
11374727 10 g 10g
3 options
This item is not returnable. View return policy

Product Code. 11384727

Brand: Thermo Scientific Alfa Aesar 039755.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used in piezoelectric devices | CAS: 12047-27-7 | BaO3Ti | 233.19 g/mol

Barium titanium oxide is used in piezoelectric devices. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12047-27-7
Molecular Formula BaO3Ti
Molecular Weight (g/mol) 233.19
MDL Number MFCD00003447
InChI Key WNKMTAQXMLAYHX-UHFFFAOYSA-N
IUPAC Name barium(2+) oxotitaniumbis(olate)
SMILES [Ba++].[O-][Ti]([O-])=O

Specifications

Physical Form Viscous Liquid
Quantity 50 g
UN Number UN1993
Solubility Information Soluble in alcohols
Concentration or Composition (by Analyte or Components) Oxide ≈35 wt%
Chemical Name or Material Barium titanium oxide, Polymeric precursor

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.