missing translation for 'onlineSavingsMsg'
Learn More

Barium titanium oxide, 99%

Product Code. 11351719
Change view
Click to view available options
Quantity:
50 g
500 g
2 kg
Unit Size:
2kg
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11351719 500 g 500g
11331719 50 g 50g
11341719 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11351719 Supplier Thermo Scientific Alfa Aesar Supplier No. 012348.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ferroelectric ceramics, single crystals, storage devices, dielectric amplifiers | CAS: 12047-27-7 | BaO3Ti | 233.19 g/mol

Barium titanium oxide is a dielectric ceramic used for capacitors. It is a piezoelectric material for microphones and other transducers. Finds applications in nonlinear optics and photorefractive applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12047-27-7
Molecular Formula BaO3Ti
Molecular Weight (g/mol) 233.19
MDL Number MFCD00003447
InChI Key WNKMTAQXMLAYHX-UHFFFAOYSA-N
Synonym Barium titanate
IUPAC Name barium(2+) oxotitaniumbis(olate)
SMILES [Ba++].[O-][Ti]([O-])=O

Specifications

Melting Point 1620°C
Physical Form Powder
Quantity 500 g
Merck Index 14,1000
Solubility Information Insoluble in water, alkalies; Slightly soluble in dilute acids; Soluble in concentrated acids
Formula Weight 233.19
Percent Purity 99%
Odor Odorless
Density 5.85 g/mL
Assay (metals basis)
Chemical Name or Material Barium titanium oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.