missing translation for 'onlineSavingsMsg'
Learn More

Barium titanium(IV) 2-ethylhexanoate pentaisopropoxide, 99.5+% (metals basis), produces 5% BaTiO3, in isopropanol, Thermo Scientific Chemicals

Product Code. 11324928
Click to view available options
Quantity:
25 mL
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 336169-47-2
Molecular Formula C23H50BaO7Ti
MDL Number MFCD01310249
Synonym Barium titanium(IV) 2-ethylhexano-isopropoxide

Specifications

Quantity 25 mL
UN Number UN1219
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 623.89
Percent Purity ≥99.5%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Barium titanium (IV) 2-ethylhexanoate pentaisopropoxide, Produces 5% BaTiO3, in isopropanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'