missing translation for 'onlineSavingsMsg'
Learn More

Barium titanium(IV) 2-ethylhexanoate pentaisopropoxide, 99.5+% (metals basis), produces 5% BaTiO3, in isopropanol, Thermo Scientific Chemicals

Product Code. 11440630
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
This item is not returnable. View return policy

Product Code. 11440630

Brand: Thermo Scientific Alfa Aesar 043893.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 336169-47-2
Molecular Formula C23H50BaO7Ti
MDL Number MFCD01310249
Synonym Barium titanium(IV) 2-ethylhexano-isopropoxide

Specifications

Quantity 500 mL
UN Number UN1219
Sensitivity Moisture Sensitive
Formula Weight 623.89
Percent Purity ≥99.5%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Barium titanium (IV) 2-ethylhexanoate pentaisopropoxide, Produces 5% BaTiO3, in isopropanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.