missing translation for 'onlineSavingsMsg'
Learn More

Barium titanium(IV) 2-ethylhexanoate pentaisopropoxide, 99.5+% (metals basis), produces 5% BaTiO3, in isopropanol, Thermo Scientific Chemicals

Product Code. 11334928
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334928 100 mL 100mL
11324928 25 mL 25mL
11440630 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334928 Supplier Thermo Scientific Alfa Aesar Supplier No. 043893.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 336169-47-2
Molecular Formula C23H50BaO7Ti
MDL Number MFCD01310249
Synonym Barium titanium(IV) 2-ethylhexano-isopropoxide

Specifications

Quantity 100 mL
UN Number UN1219
Sensitivity Moisture Sensitive
Formula Weight 623.89
Percent Purity ≥99.5%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Barium titanium (IV) 2-ethylhexanoate pentaisopropoxide, Produces 5% BaTiO3, in isopropanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.