missing translation for 'onlineSavingsMsg'
Learn More

Barium thiosulfate, 98%

Product Code. 11407197
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11407197 25 g 25g
11417197 100 g 100g
11427197 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11407197 Supplier Thermo Scientific Alfa Aesar Supplier No. A14981.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 35112-53-9 | BaO3S2 | 249.444 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35112-53-9
Molecular Formula BaO3S2
Molecular Weight (g/mol) 249.444
MDL Number MFCD00040661
InChI Key ONPIOWQPHWNPOQ-UHFFFAOYSA-L
Synonym barium thiosulfate, unii-gpa80950ab, barium 2+ hypo, barium thiosulphate, barium 2+ thiosulphate, acmc-20alqo, ba.so32, thiosulfuric acid h2s2o3 , barium salt 1:1, barium thiosulfate h2o, barium 2+ ion hypo
PubChem CID 169663
IUPAC Name barium(2+);dioxido-oxo-sulfanylidene-$l^{6}-sulfane
SMILES [O-]S(=O)(=S)[O-].[Ba+2]

Specifications

Quantity 25 g
Linear Formula BaS2O3
UN Number UN1564
Formula Weight 249.46
Percent Purity 98%
Density 3.45
Chemical Name or Material Barium thiosulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.