missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Barium thiocyanate trihydrate, 98%
Click to view available options
Quantity:
50g
Specifications
CAS | 68016-36-4 |
Density | 2.2 |
Molecular Formula | Ba(SCN)2·3H2O |
MDL Number | MFCD00150182 |
Quantity | 50g |
UN Number | UN1564 |
Sensitivity | Light sensitive |
Synonym | c2ban2s2.3h2o, cyanosulfanyl bario sulfanylcarbonitrile trihydrate |
Formula Weight | 307.55 (253.49 Anhydrous) |
Percent Purity | 98% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'