missing translation for 'onlineSavingsMsg'
Learn More

Barium tartrate, 98%

Product Code. 11434157
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11434157

Brand: Thermo Scientific Alfa Aesar A19604.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5908-81-6 | C4H4BaO6 | 285.397 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5908-81-6
Molecular Formula C4H4BaO6
Molecular Weight (g/mol) 285.397
MDL Number MFCD00054437
InChI Key HQYOWPDUMCBSLQ-UHFFFAOYSA-L
Synonym barium tartrate, barium 2+ tartrate, acmc-20alqr, barium 2+ ion tartrate, barium 2+ ; 2,3-dihydroxybutanedioate
PubChem CID 6453753
IUPAC Name barium(2+);2,3-dihydroxybutanedioate
SMILES C(C(C(=O)[O-])O)(C(=O)[O-])O.[Ba+2]

Specifications

Quantity 250 g
UN Number UN1564
Beilstein 6120238
Solubility Information Very slightly soluble in water
Formula Weight 285.41
Percent Purity 98%
Odor Odorless
Density 2.98
Chemical Name or Material Barium tartrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.