missing translation for 'onlineSavingsMsg'
Learn More

Barium sulfate, 98%, extra pure, Thermo Scientific Chemicals

Catalog Number 10442031
20.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7727-43-7
BaO4S
233.38
MFCD00003455
TZCXTZWJZNENPQ-UHFFFAOYSA-L
barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
24414
[Ba++].[O-]S([O-])(=O)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7727-43-7
BaO4S
233.38
MFCD00003455
TZCXTZWJZNENPQ-UHFFFAOYSA-L
barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
24414
[Ba++].[O-]S([O-])(=O)=O

Barium sulfate, 98%, BaO4S, CAS Number-7727-43-7, barite, baritop, barium sulfate, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast, 25kg, Passes test, < 0.3%, 97.5 to 100.5%, White, 4.5000g/mL, 231-784-4, 233.4, Extra Pure, 10ppm max., TZCXTZWJZNENPQ-UHFFFAOYSA-L

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1580.0°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 97.5 to 100.5%
Linear Formula BaSO4
Merck Index 15,99
Solubility Information Solubility in water: 0.0022g/L (18°C). Other solubilities: practically insoluble in diluted acids, soluble in hot conc. sulfuric acid, insoluble in ethanol
IUPAC Name barium(2+);sulfate
Formula Weight 233.4
Percent Purity 98%
Grade Extra Pure
Loss on Ignition 2.0% (600°C) max.
Packaging Plastic bottle
Specific Gravity 4.5
Density 4.5000g/mL
Chemical Name or Material Barium sulfate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'