missing translation for 'onlineSavingsMsg'
Learn More

Barium sulfate, 98%, extra pure

Product Code. 10002890 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Unit Size:
1kg
25kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10002890 1 kg Plastic bottle 1kg
10442031 500 g Plastic bottle 500g
11934241 5 kg Plastic bottle 5kg
15109253 25 kg Fibre drum 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10002890 Supplier Thermo Scientific Acros Supplier No. 222510010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Barium sulfate, 98%, BaO4S, CAS Number-7727-43-7, barite, baritop, barium sulfate, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast, 25kg, Passes test, < 0.3%, 97.5 to 100.5%, White, 4.5000g/mL, 231-784-4, 233.4, Extra Pure, 10ppm max., TZCXTZWJZNENPQ-UHFFFAOYSA-L | CAS: 7727-43-7 | BaO4S | 233.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-43-7
Molecular Formula BaO4S
Molecular Weight (g/mol) 233.38
MDL Number MFCD00003455
InChI Key TZCXTZWJZNENPQ-UHFFFAOYSA-L
Synonym barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
PubChem CID 24414
SMILES [Ba++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1580.0°C
Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range 97.5 to 100.5%
Linear Formula BaSO4
Merck Index 15,99
Solubility Information Solubility in water: 0.0022g/L (18°C). Other solubilities: practically insoluble in diluted acids, soluble in hot conc. sulfuric acid, insoluble in ethanol
IUPAC Name barium(2+) sulfate
Formula Weight 233.4
Percent Purity 98%
Grade Extra Pure
Loss on Ignition 2.0% (600°C) max.
Packaging Plastic bottle
Specific Gravity 4.5
Density 4.5000g/mL
Chemical Name or Material Barium sulfate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.