missing translation for 'onlineSavingsMsg'
Learn More

Barium Sulfate, 99%, ReagentPlus™, Solstice

Product Code. 15692600 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15692600 500 g Poly Bottle 500g
15672600 100 g Poly Bottle 100g
15682600 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15692600 Supplier Honeywell Research Chemicals Supplier No. 243353500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-43-7
Molecular Formula BaO4S
Molecular Weight (g/mol) 233.38
MDL Number MFCD00003455
InChI Key TZCXTZWJZNENPQ-UHFFFAOYSA-L
Synonym barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
PubChem CID 24414
SMILES [Ba++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1580°C
Color White
Quantity 500 g
Linear Formula BaSO4
UN Number NONH for all modes of transport
IUPAC Name barium(2+);sulfate
Formula Weight 233.39g/mol
Percent Purity 99%
Grade ReagentPlus
Packaging Poly Bottle
Appearance Powder
Density 4.4g/cm3
Chemical Name or Material Barium Sulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.