missing translation for 'onlineSavingsMsg'
Learn More

Barium silicon oxide, Thermo Scientific Chemicals

Codice prodotto. 11357098
Cambia vista
Click to view available options
Quantity:
100 g
500 g
Dimensione della confezione:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Dimensione della confezione
11357098 500 g 500g
11367098 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11357098 Fornitore Thermo Scientific Alfa Aesar N. del fornitore 015122.36
 Restricted Item
Estimated Shipment: 29-09-2026
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 13255-26-0 | Molecular Formula: BaO3Si | Molecular Weight: 213.409 g/mol

Barium silicate films that are suitable for use as transmission media in integrated optical circuits. It is shown that the films, which can be produced with a wide range of refractive indices by suitable selection of the ratio of the target constituents, exhibit low optical attenuation. Hence it is used for integrated optical circuits. It is used in ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 13255-26-0
Molecular Formula BaO3Si
Molecular Weight (g/mol) 213.409
MDL Number MFCD00014186
InChI Key HMOQPOVBDRFNIU-UHFFFAOYSA-N
Synonym barium silicate, barium metasilicate, silicic acid, barium salt, barium silicon oxide, barium mesosilicate, barium disilicate, bariumsiliconoxide, ba.so3i, barium meta silicate, silicic acid h2sio3 , barium salt 1:1
PubChem CID 166725
IUPAC Name barium(2+);dioxido(oxo)silane
SMILES [O-][Si](=O)[O-].[Ba+2]

Specifica

Melting Point 1604°C
Color White
Physical Form Powder
Quantity 500 g
Solubility Information Insoluble in water. Soluble in acids.
Formula Weight 213.4
Refractive Index 1.67
Density 4.399
Chemical Name or Material Barium silicon oxide

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.