missing translation for 'onlineSavingsMsg'
Learn More

Barium peroxide, ≥95% peroxide (as BaO2) basis, powder, Honeywell Fluka™

Product Code. 15691040 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
50 kg
Packaging:
Fiberboard Box
Poly Bottle
Unit Size:
500g
50kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15691040 500 g Poly Bottle 500g
15601050 50 kg Fiberboard Box 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15691040 Supplier Honeywell-Fluka Supplier No. 11428500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

≥95% peroxide (as BaO2) basis, powder

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1304-29-6
Molecular Formula BaO2
Molecular Weight (g/mol) 169.325
MDL Number MFCD00003454
InChI Key ZJRXSAYFZMGQFP-UHFFFAOYSA-N
Synonym barium peroxide, barium dioxide, barium binoxide, barium superoxide, barium oxide, per, bario perossido di, bariumperoxid german, bariumperoxyde dutch, dioxyde de baryum french, barium peroxide ba o2
PubChem CID 14773
IUPAC Name barium(2+);peroxide
SMILES [O-][O-].[Ba+2]

Specifications

Melting Point 450°C
Color Brown
Physical Form Powder
Quantity 500 g
UN Number UN1449
Formula Weight 169.33g/mol
Percent Purity ≥95% (peroxide (as BaO2) basis)
Packaging Poly Bottle
Appearance Powder
Density 4.960g/cu.cm
Chemical Name or Material Barium Peroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.