missing translation for 'onlineSavingsMsg'
Learn More

Barium perchlorate trihydrate, Reagent Grade

Product Code. 11341768
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11341768 250 g 250g
11351768 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11341768 Supplier Thermo Scientific Alfa Aesar Supplier No. 011656.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10294-39-0 | BaCl2H6O11 | 390.264 g/mol

Barium perchlorate, trihydrate, reagent is an extremely powerful oxidizing agent, and therefore commonly used in industry and pyrotechnics. It is also used in RPE-For analysis. It is used as an oxidizing agent, for gas drying, for ribonuclease determination and in the water analysis for sulfate titration with Thorin as indicator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-39-0
Molecular Formula BaCl2H6O11
Molecular Weight (g/mol) 390.264
MDL Number MFCD00149148
InChI Key NBHDMFAHVJWOHD-UHFFFAOYSA-L
Synonym barium perchlorate trihydrate, barium perchlorate trihydrate, reagent grade, ba.2clo4.3h2o, acmc-1bt65, barium perchlorate-water 1/2/3, barium 2+ trihydrate diperchlorate
PubChem CID 53249218
IUPAC Name barium(2+);diperchlorate;trihydrate
SMILES O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2]

Specifications

Physical Form Crystalline
Quantity 250 g
UN Number UN1447
Merck Index 14,987
Solubility Information Soluble in water, methanol; Slightly soluble in ethanol, ethyl acetate, acetone; Insoluble in ether
Formula Weight 390.29 (336.24 Anhydrous)
Grade Reagent
Refractive Index 1.533
Density 2.74 g/mL
Chemical Name or Material Barium perchlorate trihydrate
compliance-icons compliance-icons
Product Identifier
  • Barium perchlorate trihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Oxidising solid Category 1
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Barium perchlorate trihydrate

RUO – Research Use Only

Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.