missing translation for 'onlineSavingsMsg'
Learn More

Barium perchlorate trihydrate, 99+%, Thermo Scientific™

Product Code. 15463157
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15463157 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15463157 Supplier Thermo Scientific Alfa Aesar Supplier No. A17598.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 10294-39-0
Quantity 100 g
Molecular Formula BaCl2H6O11
MDL Number MFCD00149148
UN Number UN1447
Merck Index 14,987
Synonym barium perchlorate trihydrate, barium perchlorate trihydrate, reagent grade, ba.2clo4.3h2o, acmc-1bt65, barium perchlorate-water 1/2/3, barium 2+ trihydrate diperchlorate
InChI Key NBHDMFAHVJWOHD-UHFFFAOYSA-L
SMILES O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2]
IUPAC Name barium(2+);diperchlorate;trihydrate
Molecular Weight (g/mol) 390.264
PubChem CID 53249218
Formula Weight 390.29 (336.24 Anhydrous)
Percent Purity ≥99%
Odor Odorless
Chemical Name or Material Barium perchlorate trihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.