missing translation for 'onlineSavingsMsg'
Learn More

Barium perchlorate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals

£35.60 - £124.00

Chemical Identifiers

CAS 13465-95-7
Molecular Formula BaCl2O8
Molecular Weight (g/mol) 336.23
MDL Number MFCD00003440
InChI Key OOULUYZFLXDWDQ-UHFFFAOYSA-L
Synonym barium perchlorate, barium diperchlorate, perchloric acid, barium salt, unii-ey48pa0c98, barium 2+ diperchlorate, barium 2+ ion diperchlorate, barium perchlorate, anhydrous, perchloric acid, bariumsalt 2:1, perchloric acid, barium salt 2:1, acmc-20alqv
PubChem CID 61623
IUPAC Name barium(2+);diperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10720731
View Documents Promotion Details
Thermo Scientific Acros
221271000
100 g Glass bottle
£35.60
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10730731
View Documents Promotion Details
Thermo Scientific Acros
221275000
500 g Glass bottle
£124.00
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

13465-95-7
336.23
OOULUYZFLXDWDQ-UHFFFAOYSA-L
61623
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2]
BaCl2O8
MFCD00003440
barium perchlorate, barium diperchlorate, perchloric acid, barium salt, unii-ey48pa0c98, barium 2+ diperchlorate, barium 2+ ion diperchlorate, barium perchlorate, anhydrous, perchloric acid, bariumsalt 2:1, perchloric acid, barium salt 2:1, acmc-20alqv
barium(2+);diperchlorate

Specifications

505.0°C
White
98.5% min. (Complexometry)
Ba(ClO4)2
15, 983
Solubility in water: soluble. Other solubilities: soluble in methanol, slightly soluble in ethanol, ethyl acetate and, acetone
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2]
336.23
336.23
Extra Pure
3.2000g/mL
13465-95-7
Powder
BaCl2O8
MFCD00003440
barium perchlorate, barium diperchlorate, perchloric acid, barium salt, unii-ey48pa0c98, barium 2+ diperchlorate, barium 2+ ion diperchlorate, barium perchlorate, anhydrous, perchloric acid, bariumsalt 2:1, perchloric acid, barium salt 2:1, acmc-20alqv
OOULUYZFLXDWDQ-UHFFFAOYSA-L
barium(2+);diperchlorate
61623
99%
3.2
Barium perchlorate, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
May cause fire or explosion; strong oxidizer.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep/Store away from clothing/other combustible materials.
Take any precaution to avoid mixing with combustibles.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 236-710-4

missing translation for 'rtecsNumber' : SC7550000

missing translation for 'tsca' : TSCA

Product Identifier
  • Barium perchlorate, anhydrous

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Oxidising solid Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H271-May cause fire or explosion; strong oxidiser.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only