missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium oxide, 90%, extra pure, powder, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 2000.0°C |
Melting Point | 1920.0°C |
Color | Gray to Green-Yellow |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 90% |
Linear Formula | BaO |
Fieser | 01,683; 09,23 |
Merck Index | 15,982 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in diluted acids, alcohols, insoluble in acetone, nh3, slowly but considerably soluble in methanol,, ethanol forming barium alcoholate |
Show More |
Safety and Handling
Product Identifier
- Barium oxide
- Danger
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H301-Toxic if swallowed.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- MIXTURE LIST-Contains : Barium oxide, bariumcarbonate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'