missing translation for 'onlineSavingsMsg'
Learn More

Barium oxide, 90%, extra pure, powder

Product Code. 10104600 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
2.5kg
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10104600 500 g Plastic bottle 500g
10424555 25 g Glass Bottle 25g
11994131 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10104600 Supplier Thermo Scientific Acros Supplier No. 207835000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1304-28-5 | BaO | 153.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1304-28-5 , 513-77-9 , 1304-29-6
Molecular Formula BaO
Molecular Weight (g/mol) 153.33
MDL Number MFCD00003453
InChI Key QVQLCTNNEUAWMS-UHFFFAOYSA-N
Synonym barium oxide, baryta, barium monoxide, calcined baryta, barium protoxide, oxyde de baryum french, barium oxide, obtained by calcining witherite, oxyde de baryum
PubChem CID 62392
IUPAC Name oxobarium
SMILES O=[Ba]

Specifications

Boiling Point 2000.0°C
Melting Point 1920.0°C
Color Gray to Green-Yellow
Physical Form Powder
Quantity 500 g
Assay Percent Range 90%
Linear Formula BaO
Fieser 01,683; 09,23
Merck Index 15,982
Solubility Information Solubility in water: soluble. Other solubilities: soluble in diluted acids, alcohols, insoluble in acetone, nh3, slowly but considerably soluble in methanol,, ethanol forming barium alcoholate
Formula Weight 153.34
Percent Purity 90%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Barium oxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Barium oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H332-Harmful if inhaled.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.