missing translation for 'onlineSavingsMsg'
Learn More

Barium oxalate, Puratronic™, 99.999% (metals basis)

Product Code. 11352108
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11352108

Brand: Thermo Scientific Alfa Aesar 013009.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 516-02-9 | C2BaO4 | 225.345 g/mol

Barium oxalate, Puratronic isused as a green pyrotechnic colorant generally in specialized pyrotechnic compositions containing magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 516-02-9
Molecular Formula C2BaO4
Molecular Weight (g/mol) 225.345
MDL Number MFCD00054429
InChI Key GXUARMXARIJAFV-UHFFFAOYSA-L
Synonym barium oxalate, unii-54r8vvf8zk, ethanedioic acid, barium salt 1:1, barium 2+ ; oxalate, barium 2+ ion oxalate, 54r8vvf8zk, oxalic acid barium, barium 2+ oxalate ion, acmc-1b1e7, ethanedioic acid,barium salt 1:1
PubChem CID 68201
IUPAC Name barium(2+);oxalate
SMILES C(=O)(C(=O)[O-])[O-].[Ba+2]

Specifications

Melting Point 400°C (decomposition)
Color White
Physical Form Powder
Quantity 5 g
Linear Formula BaC2O4
UN Number UN1564
Merck Index 14,985
Solubility Information Soluble in water.
Formula Weight 225.36
Percent Purity 99.999%
Grade Puratronic
Odor Odorless
Density 2.658 g/mL
Assay (metals basis)
Chemical Name or Material Barium oxalate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.