missing translation for 'onlineSavingsMsg'
Learn More

Barium oxalate, Puratronic™, 99.999% (metals basis)

Product Code. 11342108
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342108 100 g 100g
11352108 5 g 5g
11362108 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11342108 Supplier Thermo Scientific Alfa Aesar Supplier No. 013009.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 516-02-9 | C2BaO4 | 225.345 g/mol

Barium oxalate, Puratronic isused as a green pyrotechnic colorant generally in specialized pyrotechnic compositions containing magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 516-02-9
Molecular Formula C2BaO4
Molecular Weight (g/mol) 225.345
MDL Number MFCD00054429
InChI Key GXUARMXARIJAFV-UHFFFAOYSA-L
Synonym barium oxalate, unii-54r8vvf8zk, ethanedioic acid, barium salt 1:1, barium 2+ ; oxalate, barium 2+ ion oxalate, 54r8vvf8zk, oxalic acid barium, barium 2+ oxalate ion, acmc-1b1e7, ethanedioic acid,barium salt 1:1
PubChem CID 68201
IUPAC Name barium(2+);oxalate
SMILES C(=O)(C(=O)[O-])[O-].[Ba+2]

Specifications

Melting Point 400°C (decomposition)
Color White
Physical Form Powder
Quantity 100 g
Linear Formula BaC2O4
UN Number UN1564
Merck Index 14,985
Solubility Information Soluble in water.
Formula Weight 225.36
Percent Purity 99.999%
Grade Puratronic
Odor Odorless
Density 2.658 g/mL
Assay (metals basis)
Chemical Name or Material Barium oxalate
Show More Show Less
compliance-icons
Product Identifier
  • Barium oxalate, Puratronic
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Ethanedioic acid, barium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.