missing translation for 'onlineSavingsMsg'
Learn More

Barium nitrate, Puratronic™, 99.999% (metals basis)

Product Code. 11310859
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 11310859

Brand: Thermo Scientific Alfa Aesar 010648.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10022-31-8 | BaN2O6 | 261.34 g/mol

Barium nitrate is used by the oil and gas industries to make drilling muds. It is used to make  paints, bricks, tiles, glass, rubber, and other barium compounds. It also used in is used in signal flares and water softener. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10022-31-8
Molecular Formula BaN2O6
Molecular Weight (g/mol) 261.34
MDL Number MFCD00003442
InChI Key IWOUKMZUPDVPGQ-UHFFFAOYSA-N
Synonym barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2
PubChem CID 24798
SMILES [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 592°C (decomposition)
Color White
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in water. Slightly soluble in ethanol and acetone. Partially soluble in cold water.
IUPAC Name barium(2+);dinitrate
Formula Weight 261.35
Sensitivity Hygroscopic
Appearance White crystalline
Density 3.24
Chemical Name or Material Barium nitrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.