missing translation for 'onlineSavingsMsg'
Learn More

Barium nitrate, ACS, 99+%

Product Code. 11366588
Change view
Click to view available options
Quantity:
50 g
500 g
2 kg
Unit Size:
2kg
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11366588 2 kg 2kg
11386588 50 g 50g
11376588 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11366588 Supplier Thermo Scientific Alfa Aesar Supplier No. 012194.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10022-31-8 | BaN2O6 | 261.34 g/mol

Barium nitrate is used in the manufacture of special glasses, in fire works industries and for green-fire in pyrotechnics. It is widely used in defense applications. It is a precursor material for barium oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10022-31-8
Molecular Formula BaN2O6
Molecular Weight (g/mol) 261.34
MDL Number MFCD00003442
InChI Key IWOUKMZUPDVPGQ-UHFFFAOYSA-N
Synonym barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2
PubChem CID 24798
SMILES [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 592°C (decomposition)
Color White
Physical Form Crystalline
Quantity 2 kg
Assay Percent Range >99%
Solubility Information Soluble in water. Insoluble in alcohol.
IUPAC Name barium(2+);dinitrate
Formula Weight 261.35
Grade ACS
Sensitivity Hygroscopic
Appearance White crystalline
Density 3.24
Chemical Name or Material Barium nitrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.