missing translation for 'onlineSavingsMsg'
Learn More

Barium molybdenum oxide, 99.9% (metals basis excluding Sr), Thermo Scientific Chemicals

Product Code. 11389347
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389347 25 g 25g
11379347 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389347 Supplier Thermo Scientific Alfa Aesar Supplier No. 040205.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Electronic and optical equipment, pigment in paints and other protective coatings

Barium molybdenum oxide is used in electronic and optical equipment, pigment in paints and other protective coatings. It is also used as enamel of the adhesive products for iron's adhesion strong.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-37-3
MDL Number MFCD00014182
Synonym Barium molybdate

Specifications

Melting Point ∼1600°C
Color White
Physical Form Powder
Quantity 25 g
UN Number UN1564
Solubility Information Slightly soluble in water and acids
Formula Weight 297.28
Percent Purity 99.9%
Odor Odorless
Density 4.98g/mL
Assay (metals basis excluding Sr)
Chemical Name or Material Barium molybdenum oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.