missing translation for 'onlineSavingsMsg'
Learn More

Barium metaphosphate, 90+%, Thermo Scientific Chemicals

Product Code. 11388067
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388067 100 g 100g
11378067 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388067 Supplier Thermo Scientific Alfa Aesar Supplier No. 022641.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Barium metaphosphate, used in glasses, porcelains, and enamels, as a flame retardant and in phosphors applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13762-83-9
Molecular Formula BaO6P2
Molecular Weight (g/mol) 295.27
MDL Number MFCD00061417
InChI Key RYSCSJQMVDRYDV-UHFFFAOYSA-J
Synonym barium metaphosphate, barium 2+ ; dioxido oxo phosphanium, ba.2o3p, bismetaphosphoric acid barium salt, barium 2+ bis trioxidophosphate 1-
PubChem CID 16164194
SMILES [Ba+4].[O-][P]([O-])=O.[O-][P]([O-])=O

Specifications

Melting Point 1560°C
Physical Form Powder
Quantity 100 g
UN Number UN1564
Solubility Information Insoluble in water; Soluble in acids by a slow dissolution
Formula Weight 295.25
Percent Purity ≥90%
Odor Odorless
Chemical Name or Material Barium metaphosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.