missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium metaphosphate, 90+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Barium metaphosphate, used in glasses, porcelains, and enamels, as a flame retardant and in phosphors applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1560°C |
Physical Form | Powder |
Quantity | 500 g |
UN Number | UN1564 |
Solubility Information | Insoluble in water; Soluble in acids by a slow dissolution |
Formula Weight | 295.25 |
Percent Purity | ≥90% |
Odor | Odorless |
Chemical Name or Material | Barium metaphosphate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'